Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MANSC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001283723.1 | NP_001270652.1 | 2 - 3 | 470 | 100 | |
XM_005570174.2 | XP_005570231.1 | 2 - 3 | 687 | 100 | ||
XM_015430178.1 | XP_015285664.1 | 2 - 3 | 489 | 100 | ||
XM_015430179.1 | XP_015285665.1 | 2 - 3 | 395 | 100 | ||
GenBank mRNA | AB060888.1 | - | 2 - 3 | 470 | 100 |