Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SRD5A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001283467.1 | NP_001270396.1 | 4 - 5 | 710 | 82 | |
GenBank mRNA | S77165.1 | - | 4 - 5 | 710 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Probe spans exons |