Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC2A12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001284710.1 | NP_001271639.1 | 1 - 2 | 273 | 91 | |
GenBank mRNA | AB056798.1 | - | 1 - 2 | 273 | 91 |