Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: APOL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001284822.1 | NP_001271751.1 | 3 - 4 | 358 | 99 | |
XM_015456953.1 | XP_015312439.1 | 3 - 4 | 514 | 99 | ||
XM_015456954.1 | XP_015312440.1 | 3 - 4 | 796 | 99 | ||
GenBank mRNA | AB173502.1 | - | 3 - 4 | 358 | 99 | |
AB220574.1 | - | 3 - 4 | 357 | 99 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Probe spans exons |