Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CGAS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015449182.1 | XP_015304668.1 | 3 - 4 | 1335 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Probe spans exons |