Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBXL15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005566299.2 | XP_005566356.1 | 3 - 4 | 832 | 50 | |
XM_015456615.1 | XP_015312101.1 | 3 - 4 | 397 | 50 | ||
XM_015456616.1 | XP_015312102.1 | 3 - 4 | 871 | 50 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Probe spans exons |