Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UTP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005555132.1 | XP_005555189.1 | 1 | 1519 | 61 | |
GenBank mRNA | AB060925.1 | - | 1592 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Both primers and probe map within a single exon |