Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LUZP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005544488.2 | XP_005544545.1 | 3 - 4 | 3440 | 72 | |
XM_005544489.2 | XP_005544546.1 | 3 - 4 | 3646 | 72 | ||
XM_015441020.1 | XP_015296506.1 | 4 - 5 | 3459 | 72 | ||
XM_015441022.1 | XP_015296508.1 | 4 - 5 | 3506 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Probe spans exons |