Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC102141127 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001319498.1 | NP_001306427.1 | 2 - 3 | 412 | 84 | |
GenBank mRNA | AB059654.1 | - | 2 - 3 | 412 | 84 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Probe spans exons |