Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC102140483 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005590238.2 | XP_005590295.1 | 3 - 4 | 307 | 91 | |
XM_005590239.2 | XP_005590296.1 | 3 - 4 | 318 | 91 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Probe spans exons |