Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZFP41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005564213.2 | XP_005564270.1 | 3 - 4 | 1964 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Probe spans exons |