Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC102137934 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005544749.2 | XP_005544806.1 | 1 | 349 | 129 | |
XM_005544750.2 | XP_005544807.1 | 1 | 349 | 129 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Both primers and probe map within a single exon |