Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163432.1 | NP_001156904.1 | 3 - 4 | 700 | 62 | |
NM_007712.3 | NP_031738.2 | 3 - 4 | 703 | 62 | ||
GenBank mRNA | AF033564.1 | - | 3 - 4 | 425 | 62 | |
BC015080.1 | - | 3 - 4 | 688 | 3995512 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |