Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Edn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007902.2 | NP_031928.2 | 1 - 2 | 145 | 119 | |
GenBank mRNA | BC037042.1 | - | 1 - 2 | 374 | 4512195 | 119 |
X59556.1 | - | 1 - 2 | 136 | 119 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |