Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abcb4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008830.2 | NP_032856.2 | 24 - 25 | 3268 | 90 | |
GenBank mRNA | BC150687.1 | - | 24 - 25 | 3138 | 9087598 | 90 |
CB951185.1 | - | 1 - 2 | 66 | 90 | ||
J03398.1 | - | 24 - 25 | 3268 | 90 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Transporter Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |