Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptch1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008957.2 | NP_032983.1 | 12 - 13 | 1683 | 113 | |
GenBank mRNA | AK138524.1 | - | 5 - 6 | 527 | 113 | |
AK147626.1 | - | 12 - 13 | 1892 | 113 | ||
U46155.1 | - | 12 - 13 | 1683 | 113 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |