Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abcd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008992.2 | NP_033018.2 | 3 - 4 | 437 | 68 | |
GenBank mRNA | AJ001166.1 | - | 3 - 4 | 341 | 68 | |
AK028420.1 | - | 3 - 4 | 342 | 68 | ||
BC050102.1 | - | 3 - 4 | 545 | 30012556 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |