Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pex5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277805.1 | NP_001264734.1 | 7 - 8 | 758 | 61 | |
NM_008995.2 | NP_033021.2 | 7 - 8 | 721 | 61 | ||
GenBank mRNA | AK145111.1 | - | 7 - 8 | 694 | 61 | |
BC029748.1 | - | 7 - 8 | 728 | 2599159 | 61 | |
Z97018.1 | - | 6 - 7 | 646 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |