Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vav2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009500.1 | NP_033526.1 | 6 - 7 | 1003 | 106 | |
GenBank mRNA | BC053060.1 | - | 6 - 7 | 980 | 5704852 | 106 |
U37017.1 | - | 6 - 7 | 1003 | 106 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |