Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccna2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009828.2 | NP_033958.2 | 4 - 5 | 935 | 104 | |
GenBank mRNA | AK044924.1 | - | 4 - 5 | 938 | 104 | |
AK135861.1 | - | 4 - 5 | 907 | 104 | ||
AK215356.1 | - | 2 - 3 | 242 | 104 | ||
BC052730.1 | - | 4 - 5 | 883 | 6816226 | 104 | |
X75483.1 | - | 4 - 5 | 883 | 104 | ||
Z26580.1 | - | 4 - 5 | 892 | 104 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |