Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lepr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001122899.1 | NP_001116371.1 | 2 - 3 | 592 | 110 | |
NM_010704.2 | NP_034834.1 | 2 - 3 | 592 | 110 | ||
NM_146146.2 | NP_666258.2 | 2 - 3 | 592 | 110 | ||
GenBank mRNA | AB519174.1 | - | 2 - 3 | 101 | 110 | |
AB519175.1 | - | 2 - 3 | 110 | 110 | ||
AB519176.1 | - | 2 - 3 | 77 | 110 | ||
AB519177.1 | - | 2 - 3 | 81 | 110 | ||
AB519178.1 | - | 2 - 3 | 80 | 110 | ||
AK140785.1 | - | 2 - 3 | 94 | 110 | ||
AK143989.1 | - | 2 - 3 | 281 | 110 | ||
AK155891.1 | - | 2 - 3 | 132 | 110 | ||
BC082551.1 | - | 2 - 3 | 77 | 30621706 | 110 | |
U42467.1 | - | 2 - 3 | 100 | 110 | ||
U46135.1 | - | 2 - 3 | 100 | 110 | ||
U52915.1 | - | 2 - 3 | 554 | 110 | ||
U58861.1 | - | 2 - 3 | 122 | 110 | ||
U58862.1 | - | 2 - 3 | 132 | 110 | ||
U58863.1 | - | 2 - 3 | 122 | 110 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons |