Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Matk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285854.1 | NP_001272783.1 | 2 - 3 | 334 | 94 | |
NM_001285855.1 | NP_001272784.1 | 2 - 3 | 402 | 94 | ||
NM_010768.2 | NP_034898.1 | 3 - 4 | 470 | 94 | ||
GenBank mRNA | AK013606.1 | - | 2 - 3 | 151 | 94 | |
BC060086.1 | - | 2 - 3 | 291 | 94 | ||
BC111864.1 | - | 2 - 3 | 148 | 40046807 | 94 | |
D45243.1 | - | 2 - 3 | 402 | 94 | ||
L27738.1 | - | 3 - 4 | 224 | 94 | ||
U05210.1 | - | 2 - 3 | 123 | 94 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protein Kinase Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons |