Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abcb1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011075.2 | NP_035205.1 | 20 - 21 | 2624 | 70 | |
GenBank mRNA | AK030328.1 | - | 19 - 20 | 2567 | 70 | |
BC141363.1 | - | 20 - 21 | 2581 | 9055975 | 70 | |
BC150811.1 | - | 20 - 21 | 2581 | 9087722 | 70 | |
M14757.1 | - | 20 - 21 | 2584 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons |