Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clcn4-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK035804.1 | - | 3 - 4 | 310 | 66 | |
AK133759.1 | - | 3 - 4 | 336 | 66 | ||
AK191145.1 | - | 2 - 3 | 123 | 66 | ||
AK209368.1 | - | 2 - 3 | 137 | 66 | ||
BC005553.1 | - | 3 - 4 | 371 | 3586154 | 66 | |
BC110668.1 | - | 4 - 5 | 670 | 6391053 | 66 | |
DQ186662.1 | - | 2 - 3 | 168 | 66 | ||
Z49916.1 | - | 4 - 5 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons |