Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hsf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001256042.1 | NP_001242971.1 | 5 - 6 | 632 | 72 | |
NM_001256044.1 | NP_001242973.1 | 4 - 5 | 523 | 72 | ||
NM_011939.3 | NP_036069.1 | 5 - 6 | 632 | 72 | ||
NR_045689.1 | 5 - 6 | 632 | 72 | |||
GenBank mRNA | AB029349.1 | - | 5 - 6 | 564 | 72 | |
AB029350.1 | - | 5 - 6 | 559 | 72 | ||
AF160966.1 | - | 5 - 6 | 559 | 72 | ||
AK148056.1 | - | 4 - 5 | 495 | 72 | ||
BC138130.1 | - | 5 - 6 | 632 | 8861149 | 72 | |
BC145153.1 | - | 5 - 6 | 632 | 72 | ||
BC145154.1 | - | 5 - 6 | 632 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |