Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sema4g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011976.1 | NP_036106.1 | 9 - 10 | 1513 | 106 | |
GenBank mRNA | AF134918.1 | - | 9 - 10 | 1513 | 106 | |
BC049183.1 | - | 9 - 10 | 1506 | 6395010 | 106 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |