Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cfd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013459.3 | NP_038487.1 | 1 - 2 | 129 | 102 | |
GenBank mRNA | AK143817.1 | - | 1 - 2 | 73 | 102 | |
BC138778.1 | - | 1 - 2 | 129 | 8861800 | 102 | |
BC138779.1 | - | 1 - 2 | 129 | 8861801 | 102 | |
M11768.1 | - | 1 - 2 | 70 | 102 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protease Mammalian Gene Collection Mus musculus Inventoried Probe spans exons |