Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vti1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001293685.1 | NP_001280614.1 | 7 - 8 | 909 | 80 | |
NM_001293686.1 | NP_001280615.1 | 6 - 7 | 888 | 80 | ||
NM_016862.4 | NP_058558.1 | 6 - 7 | 888 | 80 | ||
GenBank mRNA | AF035209.1 | - | 6 - 7 | 595 | 80 | |
AK004751.1 | - | 6 - 7 | 584 | 80 | ||
AK028646.1 | - | 6 - 7 | 594 | 80 | ||
AK135463.1 | - | 6 - 7 | 651 | 80 | ||
BC089321.1 | - | 7 - 8 | 616 | 6839437 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Mus musculus Probe spans exons |