Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fxyd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019503.4 | NP_062376.2 | 6 - 7 | 368 | 60 | |
NM_052991.4 | NP_443717.1 | 6 - 7 | 351 | 60 | ||
NM_052992.3 | NP_443718.1 | 6 - 7 | 432 | 60 | ||
NM_194321.2 | NP_919302.1 | 6 - 7 | 415 | 60 | ||
GenBank mRNA | AA466595.1 | - | 6 - 7 | 368 | 60 | |
AF089734.1 | - | 6 - 7 | 294 | 60 | ||
AK002585.1 | - | 6 - 7 | 355 | 60 | ||
AK003504.1 | - | 2 - 3 | 150 | 60 | ||
BC024671.1 | - | 6 - 7 | 305 | 4208962 | 60 | |
BY132051.1 | - | 6 - 7 | 337 | 60 | ||
CA459152.1 | - | 6 - 7 | 373 | 60 | ||
CK793316.1 | - | 6 - 7 | 350 | 60 | ||
W16239.1 | - | 6 - 7 | 331 | 60 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Ion Channel Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |