Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Srpk3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019684.1 | NP_062658.1 | 4 - 5 | 414 | 79 | |
GenBank mRNA | AF043288.1 | - | 4 - 5 | 414 | 79 | |
AJ748651.1 | - | 3 - 4 | 238 | 79 | ||
AK004696.2 | - | 4 - 5 | 432 | 79 | ||
BC120883.1 | - | 4 - 5 | 409 | 40057934 | 79 | |
BC120884.1 | - | 4 - 5 | 409 | 40057937 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protein Kinase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |