Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Glp1r | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021332.2 | NP_067307.2 | 3 - 4 | 290 | 68 | |
GenBank mRNA | AJ001692.1 | - | 3 - 4 | 280 | 68 | |
AY293742.1 | - | 3 - 4 | 280 | 68 | ||
BC103624.1 | - | 3 - 4 | 279 | 68 | ||
BC105649.1 | - | 3 - 4 | 279 | 68 | ||
BC105650.1 | - | 3 - 4 | 279 | 68 | ||
BC139464.1 | - | 3 - 4 | 317 | 8862486 | 68 | |
BC139466.1 | - | 3 - 4 | 317 | 8862488 | 68 | |
BC145626.1 | - | 3 - 4 | 317 | 9054194 | 68 | |
DQ093397.1 | - | 3 - 4 | 290 | 68 | ||
DQ093398.1 | - | 3 - 4 | 290 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 G-protein coupled receptor Mammalian Gene Collection Mus musculus 1700 Probe spans exons |