Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ak1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021515.3 | NP_067490.1 | 1 - 2 | 115 | 82 | |
GenBank mRNA | AJ010109.1 | - | 1 - 2 | 111 | 82 | |
BC054366.1 | - | 1 - 2 | 104 | 5346232 | 82 | |
CF950379.1 | - | 1 - 2 | 115 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons |