Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rtn4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024226.4 | NP_077188.1 | 1 - 2 | 315 | 88 | |
GenBank mRNA | AF326337.1 | - | 1 - 2 | 95 | 88 | |
AY102283.1 | - | 1 - 2 | 308 | 88 | ||
BU848611.1 | - | 1 - 2 | 154 | 88 | ||
BY251875.1 | - | 1 - 2 | 315 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mus musculus Probe spans exons |