Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nrg4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_032002.2 | NP_114391.1 | 2 - 3 | 193 | 118 | |
GenBank mRNA | AF083067.2 | - | 1 - 2 | 71 | 118 | |
AK080089.1 | - | 2 - 3 | 171 | 118 | ||
BC034839.1 | - | 2 - 3 | 155 | 3372942 | 118 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |