Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sptlc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009269.2 | NP_033295.2 | 6 - 7 | 574 | 96 | |
GenBank mRNA | AF003823.1 | - | 6 - 7 | 578 | 96 | |
AK028713.1 | - | 6 - 7 | 585 | 96 | ||
AK043064.1 | - | 2 - 3 | 153 | 96 | ||
AK053207.1 | - | 6 - 7 | 579 | 96 | ||
AK079578.1 | - | 6 - 7 | 575 | 96 | ||
AK084391.1 | - | 6 - 7 | 578 | 96 | ||
AK084445.1 | - | 6 - 7 | 571 | 96 | ||
AK089030.1 | - | 6 - 7 | 578 | 96 | ||
AK161140.1 | - | 6 - 7 | 611 | 96 | ||
AK161811.1 | - | 2 - 3 | 153 | 96 | ||
BC046323.1 | - | 6 - 7 | 574 | 6391985 | 96 | |
BC068243.1 | - | 3432 | 96 | |||
X95641.1 | - | 6 - 7 | 573 | 96 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |