Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0100030135.mm.1 |
NM_001128605.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100030135.mm.1 |
NM_001128605.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100030135.mm.1 |
NM_011183.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100030135.mm.1 |
NM_011183.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100030135.mm.1 |
TC0100003664.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100030136.mm.1 |
NM_001128605.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100030136.mm.1 |
NM_001128605.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100030136.mm.1 |
NM_011183.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100030136.mm.1 |
NM_011183.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100030136.mm.1 |
TC0100003664.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100030137.mm.1 |
NM_001128605.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100030137.mm.1 |
NM_001128605.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100030137.mm.1 |
NM_011183.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100030137.mm.1 |
NM_011183.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100030137.mm.1 |
TC0100003664.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100030138.mm.1 |
NM_001128605.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100030138.mm.1 |
NM_001128605.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100030138.mm.1 |
NM_011183.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100030138.mm.1 |
NM_011183.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100030138.mm.1 |
TC0100003664.mm.1 |
MTA TC |