Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc37a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001145960.1 | NP_001139432.1 | 2 - 3 | 527 | 64 | |
NM_020258.4 | NP_064654.3 | 2 - 3 | 527 | 64 | ||
GenBank mRNA | AF121081.1 | - | 2 - 3 | 426 | 64 | |
AK028984.1 | - | 2 - 3 | 203 | 64 | ||
AK031045.1 | - | 2 - 3 | 465 | 64 | ||
AK079414.1 | - | 2 - 3 | 203 | 64 | ||
AK079823.1 | - | 2 - 3 | 528 | 64 | ||
BC038000.1 | - | 2 - 3 | 198 | 64 | ||
BC063326.1 | - | 2 - 3 | 215 | 30301217 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |