Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbms1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001141931.1 | NP_001135403.1 | 2 - 3 | 490 | 105 | |
NM_001141932.1 | NP_001135404.1 | 2 - 3 | 490 | 105 | ||
NM_020296.2 | NP_064692.2 | 2 - 3 | 683 | 105 | ||
GenBank mRNA | AB026569.1 | - | 2 - 3 | 406 | 105 | |
AK133054.1 | - | 2 - 3 | 211 | 105 | ||
AK135280.1 | - | 2 - 3 | 370 | 105 | ||
AK145897.1 | - | 2 - 3 | 492 | 105 | ||
AK147109.1 | - | 2 - 3 | 684 | 105 | ||
AK161102.1 | - | 2 - 3 | 440 | 105 | ||
AK164307.1 | - | 2 - 3 | 484 | 105 | ||
BC016501.1 | - | 2 - 3 | 671 | 4527742 | 105 | |
BC057866.1 | - | 2 - 3 | 522 | 5327143 | 105 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |