Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Med15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001040683.2 | NP_001035773.1 | 9 - 10 | 1412 | 70 | |
NM_001285884.1 | NP_001272813.1 | 10 - 11 | 1532 | 70 | ||
NM_001285886.1 | NP_001272815.1 | 9 - 10 | 1224 | 70 | ||
NM_033609.3 | NP_291087.2 | 10 - 11 | 1532 | 70 | ||
GenBank mRNA | AF328770.1 | - | 10 - 11 | 1408 | 70 | |
AK131186.1 | - | 9 - 10 | 1353 | 70 | ||
AK142224.1 | - | 10 - 11 | 1499 | 70 | ||
AK166522.1 | - | 9 - 10 | 1224 | 70 | ||
AK169904.1 | - | 10 - 11 | 1533 | 70 | ||
BC049891.1 | - | 5 - 6 | 996 | 70 | ||
BC054779.1 | - | 9 - 10 | 1362 | 6812541 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |