Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abcc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008576.3 | NP_032602.1 | 4 - 5 | 530 | 96 | |
GenBank mRNA | AF022908.1 | - | 4 - 5 | 492 | 96 | |
AK029876.1 | - | 4 - 5 | 531 | 96 | ||
AK150015.1 | - | 4 - 5 | 511 | 96 | ||
AK152781.1 | - | 4 - 5 | 511 | 96 | ||
BC090617.1 | - | 4 - 5 | 501 | 30943198 | 96 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |