Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kif17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010623.4 | NP_034753.1 | 13 - 14 | 95 | ||
GenBank mRNA | AB008867.1 | - | 13 - 14 | 95 | ||
AK122506.1 | - | 3 - 4 | 3470 | 95 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mus musculus Probe spans exons |