Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038589.2 | NP_001033678.1 | 4 - 5 | 519 | 79 | |
NM_021522.4 | NP_067497.2 | 5 - 6 | 624 | 79 | ||
GenBank mRNA | AB034633.1 | - | 5 - 6 | 403 | 79 | |
AK011322.1 | - | 5 - 6 | 560 | 79 | ||
AK029977.1 | - | 5 - 6 | 553 | 79 | ||
AK042356.1 | - | 1 - 2 | 384 | 79 | ||
AK045909.1 | - | 5 - 6 | 544 | 79 | ||
AK160215.1 | - | 5 - 6 | 558 | 79 | ||
AK170933.1 | - | 618 | 79 | |||
AK172477.1 | - | 5 - 6 | 611 | 79 | ||
BC005571.1 | - | 5 - 6 | 543 | 3157723 | 79 | |
BC050197.1 | - | 4 - 5 | 437 | 30006882 | 79 | |
CB520876.1 | - | 4 - 5 | 426 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protease Mammalian Gene Collection Mus musculus Probe spans exons |