Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prmt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AF169620.1 | - | 5 - 6 | 607 | 70 | |
AK077456.1 | - | 5 - 6 | 664 | 70 | ||
AK140200.1 | - | 5 - 6 | 690 | 70 | ||
AK144412.1 | - | 5 - 6 | 662 | 70 | ||
AK145829.1 | - | 5 - 6 | 664 | 70 | ||
AK146754.1 | - | 5 - 6 | 663 | 70 | ||
AK146966.1 | - | 5 - 6 | 671 | 70 | ||
AK159186.1 | - | 5 - 6 | 670 | 70 | ||
BC088997.1 | - | 5 - 6 | 621 | 70 | ||
BC122563.1 | - | 4 - 5 | 595 | 40107557 | 70 | |
BC125275.1 | - | 4 - 5 | 595 | 40107558 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |