Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: St8sia6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145838.1 | NP_665837.1 | 1 - 2 | 174 | 77 | |
GenBank mRNA | AB059554.1 | - | 1 - 2 | 174 | 77 | |
AK156557.1 | - | 1 - 2 | 711 | 77 | ||
AK172400.1 | - | 1 - 2 | 239 | 77 | ||
BC115912.1 | - | 1 - 2 | 129 | 40104779 | 77 | |
BC118063.1 | - | 1 - 2 | 129 | 40104780 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |