Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P91402 |
BC018382.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P91402 |
BC049231.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P91402 |
AK152289.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P91402 |
AK168912.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P91402 |
AY253297.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P91402 |
BC018382.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P91402 |
BC049231.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P91402 |
AK152289.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P91402 |
AK168912.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P91402 |
AY253297.1 |
mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
622532 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
925260 |
AF001863.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
925260 |
AF061744.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
925260 |
AK038225.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
925260 |
AK137907.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
925260 |
AK172164.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
925260 |
NM_011815.4 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
502845 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
882712 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
588713 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
603624 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
42597 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
576914 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
112875 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
804477 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
10061 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
800536 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
704293 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
930174 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
652192 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
176784 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
775830 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
9345 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
421776 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
236818 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
921336 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
15742 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
657737 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
440904 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
420667 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
107503 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
68474 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
79934 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
115776 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
18383 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P131655 |
- |
- |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P91402 |
BC018382.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P91402 |
BC049231.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P91402 |
AK152289.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P91402 |
AK168912.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P91402 |
AY253297.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P91402 |
BC018382.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P91402 |
BC049231.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P91402 |
AK152289.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P91402 |
AK168912.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P91402 |
AY253297.1 |
mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23301607 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23301621 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23301632 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23301636 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23301627 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23301639 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23301638 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23301640 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23301624 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23301615 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23301634 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23301606 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23301608 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23301611 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23301601 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23301605 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23301629 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23301612 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23301635 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23301617 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23301602 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23301631 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23301619 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P131655 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P05402013 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P05401990 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P05402023 |
- |
- |
Whole Mouse Genome G4122A |
A_51_P223961 |
BC018382.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P223961 |
BC049231.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P223961 |
AK152289.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P223961 |
AK168912.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P223961 |
AY253297.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P91402 |
BC018382.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P91402 |
BC049231.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P91402 |
AK152289.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P91402 |
AK168912.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P91402 |
AY253297.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P91402 |
BC018382.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P91402 |
BC049231.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P91402 |
AK152289.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P91402 |
AK168912.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P91402 |
AY253297.1 |
mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P107820 |
- |
- |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P137751 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P131655 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P03942831 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P03612251 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P03612249 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P03252839 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23301612 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23301484 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23301618 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23301648 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23301535 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23301475 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23301587 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23301440 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23301436 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23301452 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23301664 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23301534 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23301629 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23301519 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23301460 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23301594 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23301508 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23301455 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23301507 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23301521 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23301660 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23301491 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23301561 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23301625 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23301527 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23301579 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23301529 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23301605 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23301553 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23301627 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23301573 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23301653 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23301453 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23301597 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23301488 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23301477 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23301593 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23301433 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23301515 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23301486 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23301576 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23301611 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23301516 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23301546 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23301577 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23301430 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23301639 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23301652 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23301481 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23301566 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23301449 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23301464 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23301479 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23301571 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23301446 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23301466 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23301658 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23301498 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23301532 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23301540 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23301645 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23301501 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23301473 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23301554 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23301456 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23301617 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23301548 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23301510 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23301643 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23301539 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23301454 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23301568 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23301463 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23301558 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23301502 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23301632 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23301483 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23301550 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23301538 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23301570 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23301451 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23301590 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23301499 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23301619 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23301509 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23301489 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23301461 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23301443 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23301601 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23301588 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23301438 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23301562 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23301557 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23301651 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23301564 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23301636 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23301640 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23301638 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23301608 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23301505 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23301533 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23301578 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23301635 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23301642 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23301600 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23301656 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23301448 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23301536 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23301472 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23301520 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23301468 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23301638 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03377603 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23301656 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23301652 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23301635 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23301553 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23301658 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23301654 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23301636 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23301651 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23301554 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03986310 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23301561 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03289967 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23301634 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23301639 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23301501 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23301645 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23301507 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23301508 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03630472 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23301660 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04011510 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03289970 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23301544 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23301503 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23301502 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23301566 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23301662 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03986314 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23301650 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23301557 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23301505 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23301647 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03986312 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23301653 |
- |
- |