Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptprv | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007955.3 | NP_031981.3 | 7 - 8 | 1213 | 82 | |
GenBank mRNA | AF300701.1 | - | 6 - 7 | 690 | 82 | |
AK156112.1 | - | 7 - 8 | 1215 | 82 | ||
U36488.1 | - | 3 - 4 | 375 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protein Phosphatase Detect Genomic DNA Mus musculus Probe spans exons |