Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: H13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010376.4 | NP_034506.1 | 11 - 12 | 1156 | 67 | |
GenBank mRNA | AF483214.1 | - | 11 - 12 | 1123 | 67 | |
AF515662.1 | - | 11 - 12 | 1035 | 67 | ||
AJ345032.1 | - | 11 - 12 | 1166 | 67 | ||
AK007664.1 | - | 11 - 12 | 1138 | 67 | ||
AK012600.1 | - | 11 - 12 | 1149 | 67 | ||
AK028109.1 | - | 11 - 12 | 1125 | 67 | ||
AK050664.1 | - | 11 - 12 | 1099 | 67 | ||
AK077983.1 | - | 4 - 5 | 251 | 67 | ||
AK080966.1 | - | 11 - 12 | 1118 | 67 | ||
AK159176.1 | - | 11 - 12 | 1130 | 67 | ||
AK169922.1 | - | 11 - 12 | 1113 | 67 | ||
AK171790.1 | - | 10 - 11 | 959 | 67 | ||
BC034217.1 | - | 6 - 7 | 430 | 67 | ||
BC045195.1 | - | 7 - 8 | 512 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protease 3' Most Mammalian Gene Collection Mus musculus Probe spans exons |