Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cxcl16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023158.6 | NP_075647.3 | 2 - 3 | 742 | 128 | |
GenBank mRNA | AF277001.1 | - | 2 - 3 | 205 | 128 | |
AF301017.1 | - | 2 - 3 | 271 | 128 | ||
AK003085.1 | - | 2 - 3 | 401 | 128 | ||
AK009599.1 | - | 2 - 3 | 291 | 128 | ||
AK028875.1 | - | 2 - 3 | 744 | 128 | ||
AK030515.1 | - | 2 - 3 | 167 | 128 | ||
AK150224.1 | - | 2 - 3 | 430 | 128 | ||
AK151330.1 | - | 2 - 3 | 516 | 128 | ||
AK152102.1 | - | 2 - 3 | 516 | 128 | ||
AK152547.1 | - | 2 - 3 | 694 | 128 | ||
AK153192.1 | - | 2 - 3 | 458 | 128 | ||
BC019961.1 | - | 2 - 3 | 623 | 3988542 | 128 | |
GQ380016.1 | - | 2 - 3 | 238 | 128 | ||
GQ380017.1 | - | 2 - 3 | 238 | 128 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |