Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spns1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023712.3 | NP_076201.2 | 2 - 3 | 1163 | 71 | |
NR_045537.1 | 2 - 3 | 1163 | 71 | |||
GenBank mRNA | AF212372.1 | - | 2 - 3 | 654 | 71 | |
AK008721.1 | - | 2 - 3 | 660 | 71 | ||
AK033907.1 | - | 2 - 3 | 640 | 71 | ||
AK154353.1 | - | 2 - 3 | 653 | 71 | ||
AK166931.1 | - | 2 - 3 | 655 | 71 | ||
AK202033.1 | - | 1 - 2 | 65 | 71 | ||
BC002297.1 | - | 2 - 3 | 182 | 71 | ||
BC026854.1 | - | 2 - 3 | 624 | 4242049 | 71 | |
BC085491.1 | - | 2 - 3 | 607 | 6829415 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |