Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hmg20a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025812.2 | NP_080088.1 | 3 - 4 | 321 | 66 | |
GenBank mRNA | AK004596.1 | - | 3 - 4 | 341 | 66 | |
AK017716.1 | - | 3 - 4 | 524 | 66 | ||
AK032790.1 | - | 3 - 4 | 349 | 66 | ||
AK039222.1 | - | 3 - 4 | 343 | 66 | ||
AK134213.1 | - | 3 - 4 | 493 | 66 | ||
AL355705.1 | - | 3 - 4 | 307 | 66 | ||
BC013804.1 | - | 3 - 4 | 321 | 3602239 | 66 | |
BC068257.1 | - | 3 - 4 | 394 | 6434284 | 66 | |
DQ182735.1 | - | 3 - 4 | 321 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |